Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000639.2 | NP_000630.1 | 4 | 736 | ||
NM_001302746.1 | NP_001289675.1 | 3 | 690 | |||
GenBank mRNA | AF288573.1 | - | 3 | 554 | ||
AK314874.1 | - | 4 | 700 | |||
AY225406.1 | - | 4 | 542 | |||
AY858799.1 | - | 4 | 542 | |||
BC017502.1 | - | 4 | 629 | 4849770 | ||
D38122.1 | - | 4 | 730 | |||
KJ948655.1 | - | 4 | 699 | |||
U08137.1 | - | 4 | 627 | |||
U11821.1 | - | 4 | 606 | |||
X89102.1 | - | 4 | 699 |