Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000694.3 | NP_000685.1 | 10 | 1761 | ||
NM_001030010.2 | NP_001025181.1 | 9 | 1650 | |||
NM_001161473.2 | NP_001154945.1 | 10 | 1819 | |||
NM_001290058.1 | NP_001276987.1 | 9 | 1410 | |||
NM_001290059.1 | NP_001276988.1 | 9 | 1679 | |||
GenBank mRNA | AB209651.1 | - | 8 | 3916 | ||
BC002553.2 | - | 8 | 828 | |||
BC013584.2 | - | 10 | 1714 | 3878124 | ||
BC014168.2 | - | 4 | 745 | |||
BC033099.1 | - | 9 | 1588 | 5446816 | ||
EF411198.1 | - | 10 | 1788 | |||
U10868.1 | - | 10 | 1719 |