Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004378.2 | NP_004369.1 | 2, 3 | 361 | ||
GenBank mRNA | AB593091.1 | - | 2, 3 | 335 | ||
AL110113.1 | - | 2, 3 | 245 | |||
AU132726.1 | - | 2, 3 | 361 | |||
BC022069.1 | - | 2, 3 | 337 | 4797853 | ||
CR457017.1 | - | 2, 3 | 256 | |||
S74445.1 | - | 2, 3 | 330 |