Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVPR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000706.4 | NP_000697.1 | 2 | 3240 | ||
GenBank mRNA | BC074803.2 | - | 2 | 1289 | 30915313 | |
BC074804.2 | - | 2 | 1289 | 30915527 | ||
L25615.1 | - | 2 | 1318 |
Set Membership: |
G-protein coupled receptor |