Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000718.3 | NP_000709.1 | 39 | 5505 | ||
NM_001243812.1 | NP_001230741.1 | 39 | 5505 | |||
XM_011518991.2 | XP_011517293.1 | 40 | 5484 | |||
XM_011518992.2 | XP_011517294.1 | 40 | 5481 | |||
XM_011518993.2 | XP_011517295.1 | 39 | 5474 | |||
XM_011518994.2 | XP_011517296.1 | 38 | 5465 | |||
XM_017015115.1 | XP_016870604.1 | 40 | 5481 | |||
XM_017015116.1 | XP_016870605.1 | 39 | 5478 | |||
XM_017015117.1 | XP_016870606.1 | 39 | 5471 | |||
XM_017015118.1 | XP_016870607.1 | 39 | 5465 | |||
XM_017015120.1 | XP_016870609.1 | 38 | 5462 | |||
GenBank mRNA | AB209467.1 | - | 21 | 2455 | ||
AY429522.1 | - | 3 | 272 | |||
AY429523.1 | - | 4 | 400 | |||
M94172.1 | - | 39 | 5505 | |||
M94173.1 | - | 39 | 5505 |