Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004386.2 | NP_004377.2 | 3, 4 | 575 | ||
GenBank mRNA | AB210004.1 | - | 3, 4 | 519 | ||
AF026547.1 | - | 2, 3 | 477 | |||
CN282070.1 | - | 3, 4 | 575 |