Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103.3 | NP_001094.1 | 21 | 3034 | ||
NM_001278343.1 | NP_001265272.1 | 21 | 3034 | |||
NM_001278344.1 | NP_001265273.1 | 23 | 3231 | |||
GenBank mRNA | AB209521.1 | - | 23 | 3175 | ||
AK294906.1 | - | 19 | 2545 | |||
AK297469.1 | - | 15 | 2179 | |||
AK297618.1 | - | 14 | 2116 | |||
AK297628.1 | - | 14 | 1892 | |||
BC047901.2 | - | 21 | 2966 | 6199155 | ||
CB153006.1 | - | 1 | 121 | |||
M86406.1 | - | 21 | 2989 |