Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127.3 | NP_001118.3 | 13 | 1881 | ||
NM_001166019.1 | NP_001159491.1 | 13 | 1881 | |||
NM_145730.2 | NP_663782.2 | 13 | 1881 | |||
GenBank mRNA | AK294406.1 | - | 4 | 445 | ||
AK296567.1 | - | 2 | 225 | |||
AK299613.1 | - | 4 | 446 | |||
BC046242.1 | - | 13 | 1881 | 5929327 | ||
CT841508.1 | - | 12 | 1724 | |||
L13939.1 | - | 12 | 1740 |