Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127.3 | NP_001118.3 | 23 | 4035 | ||
NM_001166019.1 | NP_001159491.1 | 21 | 3945 | |||
NM_145730.2 | NP_663782.2 | 21 | 4005 | |||
GenBank mRNA | BX537680.1 | - | 4 | 3417 |