Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001153.4 | NP_001144.1 | 13 | 1976 | ||
NM_001320698.1 | NP_001307627.1 | 15 | 3258 | |||
NM_001320700.1 | NP_001307629.1 | 13 | 1910 | |||
NM_001320702.1 | NP_001307631.1 | 13 | 2011 | |||
XM_017003943.1 | XP_016859432.1 | 16 | 3120 | |||
XM_017003944.1 | XP_016859433.1 | 16 | 3086 | |||
GenBank mRNA | AK058017.1 | - | 13 | 1816 | ||
D78152.1 | - | 12 | 1790 | |||
M82809.1 | - | 13 | 1801 |