Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005336.2 | NP_001005336.1 | 4 | 622 | ||
NM_001288737.1 | NP_001275666.1 | 4 | 622 | |||
NM_001288738.1 | NP_001275667.1 | 4 | 622 | |||
NM_001288739.1 | NP_001275668.1 | 4 | 622 | |||
NM_004408.3 | NP_004399.2 | 4 | 622 | |||
XM_005251763.2 | XP_005251820.1 | 4 | 622 | |||
XM_005251768.2 | XP_005251825.1 | 4 | 622 | |||
XM_005251769.2 | XP_005251826.1 | 4 | 622 | |||
XM_006716992.2 | XP_006717055.1 | 4 | 622 | |||
XM_006716993.1 | XP_006717056.1 | 4 | 622 | |||
XM_011518335.2 | XP_011516637.1 | 4 | 622 | |||
XM_011518336.2 | XP_011516638.1 | 4 | 622 | |||
XM_017014369.1 | XP_016869858.1 | 4 | 622 | |||
XM_017014370.1 | XP_016869859.1 | 4 | 622 | |||
XM_017014371.1 | XP_016869860.1 | 4 | 622 | |||
XM_017014372.1 | XP_016869861.1 | 4 | 622 | |||
XM_017014373.1 | XP_016869862.1 | 4 | 622 | |||
GenBank mRNA | AB209124.1 | - | 3 | 307 | ||
AK225179.1 | - | 4 | 531 | |||
AK295101.1 | - | 4 | 415 | |||
AK296326.1 | - | 4 | 589 | |||
AK316235.1 | - | 4 | 598 | |||
BC050279.1 | - | 4 | 598 | 5284480 | ||
BC063850.1 | - | 4 | 555 | 6184100 | ||
L07807.1 | - | 4 | 572 |