Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACVR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278579.1 | NP_001265508.1 | 3 | 422 | ||
NM_001278580.1 | NP_001265509.1 | 2 | 225 | |||
NM_001616.4 | NP_001607.1 | 2 | 311 | |||
GenBank mRNA | AK301629.1 | - | 2 | 225 | ||
AK314124.1 | - | 2 | 501 | |||
BC067417.1 | - | 2 | 239 | 7002204 | ||
BC067418.1 | - | 2 | 239 | 7002205 | ||
BC069707.1 | - | 2 | 306 | 7262543 | ||
D31770.1 | - | 2 | 316 | |||
M93415.1 | - | 3 | 326 | |||
X62381.1 | - | 2 | 298 | |||
X63128.1 | - | 2 | 168 |