Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001635.3 | NP_001626.1 | 19 | 1838 | ||
NM_139316.2 | NP_647477.1 | 18 | 1712 | |||
XM_006715689.3 | XP_006715752.1 | 18 | 3375 | |||
XM_006715690.3 | XP_006715753.1 | 18 | 2157 | |||
XM_011515271.2 | XP_011513573.1 | 19 | 2283 | |||
XM_017011995.1 | XP_016867484.1 | 19 | 3501 | |||
XM_017011996.1 | XP_016867485.1 | 18 | 2463 | |||
GenBank mRNA | AF034996.1 | - | 18 | 1565 | ||
AF498092.1 | - | 7 | 893 | |||
AF498093.1 | - | 5 | 850 | |||
AF498094.1 | - | 5 | 1762 | |||
AF498095.1 | - | 6 | 649 | |||
AF498096.1 | - | 10 | 1288 | |||
AK291447.1 | - | 19 | 1642 | |||
BC034376.1 | - | 19 | 1629 | 5181100 | ||
U07616.1 | - | 19 | 1732 | |||
X81438.1 | - | 19 | 1696 |