Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001635.3 | NP_001626.1 | 19 | 1895 | ||
NM_139316.2 | NP_647477.1 | 18 | 1769 | |||
XM_006715689.3 | XP_006715752.1 | 18 | 3432 | |||
XM_006715690.3 | XP_006715753.1 | 18 | 2214 | |||
XM_011515271.2 | XP_011513573.1 | 19 | 2340 | |||
XM_017011995.1 | XP_016867484.1 | 19 | 3558 | |||
XM_017011996.1 | XP_016867485.1 | 18 | 2520 | |||
GenBank mRNA | AF034996.1 | - | 18 | 1622 | ||
AF498092.1 | - | 7 | 950 | |||
AF498096.1 | - | 10 | 1345 | |||
AK291447.1 | - | 19 | 1699 | |||
BC034376.1 | - | 19 | 1686 | 5181100 | ||
U07616.1 | - | 19 | 1789 | |||
X81438.1 | - | 19 | 1753 |