Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317384.1 | NP_001304313.1 | 2 | 355 | ||
NM_001317387.1 | NP_001304316.1 | 2 | 355 | |||
NM_001650.5 | NP_001641.1 | 2 | 355 | |||
NM_004028.4 | NP_004019.1 | 1 | 482 | |||
XM_011525942.2 | XP_011524244.1 | 2 | 397 | |||
GenBank mRNA | AB209156.1 | - | 2 | 221 | ||
AK295069.1 | - | 2 | 226 | |||
BC022286.1 | - | 2 | 220 | 4717755 | ||
BI596912.1 | - | 2 | 255 | |||
BI667387.1 | - | 2 | 273 | |||
HQ901095.1 | - | 2 | 329 | |||
KF055862.1 | - | 2 | 200 | |||
U34845.1 | - | 1 | 418 | |||
U63622.1 | - | 2 | 239 | |||
U63623.1 | - | 1 | 143 |