Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256196.1 | NP_001243125.1 | 14 | 1731 | ||
NM_001654.4 | NP_001645.1 | 14 | 1722 | |||
XM_006724529.2 | XP_006724592.1 | 14 | 1658 | |||
XM_011543906.2 | XP_011542208.1 | 14 | 3048 | |||
XM_011543908.2 | XP_011542210.1 | 14 | 3032 | |||
XM_011543909.2 | XP_011542211.1 | 9 | 1023 | |||
GenBank mRNA | AB208831.1 | - | 10 | 4789 | ||
AF052136.1 | - | 4 | 429 | |||
AK300975.1 | - | 14 | 1578 | |||
BC002466.2 | - | 14 | 1602 | 3347509 | ||
BC007514.2 | - | 14 | 1609 | 2966604 | ||
BT019864.1 | - | 13 | 1511 | |||
KU177914.1 | - | 13 | 1511 | |||
KU177915.1 | - | 13 | 1526 | |||
M13829.1 | - | 5 | 638 | |||
X04790.1 | - | 14 | 1705 |