Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001710.5 | NP_001701.2 | 11 | 1740 | ||
GenBank mRNA | AF349679.1 | - | 11 | 1462 | ||
AK130533.1 | - | 11 | 1669 | |||
AK223400.1 | - | 11 | 1588 | |||
BC004143.2 | - | 11 | 1597 | 2959705 | ||
BC007990.2 | - | 11 | 1597 | 2959706 | ||
J00126.1 | - | 5 | 448 | |||
K01566.1 | - | 10 | 1201 | |||
L15702.1 | - | 11 | 1502 | |||
S67310.1 | - | 11 | 1573 | |||
X72875.1 | - | 11 | 1462 |