Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001715.2 | NP_001706.2 | 10 | 1596 | ||
XM_011543824.1 | XP_011542126.1 | 10 | 1695 | |||
XM_011543825.2 | XP_011542127.1 | 10 | 1440 | |||
XM_011543827.1 | XP_011542129.1 | 9 | 1571 | |||
XM_017013730.1 | XP_016869219.1 | 9 | 1493 | |||
GenBank mRNA | AK313751.1 | - | 10 | 1185 | ||
BC007371.2 | - | 10 | 1159 | 3634568 | ||
BC018703.1 | - | 10 | 1460 | |||
BC032413.1 | - | 10 | 1205 | 5216030 | ||
S76617.1 | - | 10 | 1603 | |||
Z33998.1 | - | 10 | 1237 |