Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRDT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242805.2 | NP_001229734.2 | 5 | 837 | ||
NM_001242806.2 | NP_001229735.2 | 4 | 768 | |||
NM_001242807.2 | NP_001229736.2 | 3 | 436 | |||
NM_001242808.2 | NP_001229737.2 | 3 | 630 | |||
NM_001242810.2 | NP_001229739.2 | 4 | 1124 | |||
NM_001726.4 | NP_001717.3 | 4 | 563 | |||
NM_207189.3 | NP_997072.2 | 4 | 768 | |||
XM_006710853.3 | XP_006710916.1 | 5 | 1353 | |||
XM_006710854.3 | XP_006710917.1 | 6 | 1198 | |||
XM_006710855.3 | XP_006710918.1 | 6 | 1203 | |||
XM_006710856.3 | XP_006710919.1 | 6 | 1177 | |||
XM_006710857.3 | XP_006710920.1 | 6 | 1167 | |||
XM_011542032.2 | XP_011540334.1 | 6 | 687 | |||
XM_011542033.2 | XP_011540335.1 | 6 | 661 | |||
XM_011542034.2 | XP_011540336.1 | 6 | 677 | |||
XM_011542035.2 | XP_011540337.1 | 6 | 651 | |||
XM_011542036.2 | XP_011540338.1 | 4 | 566 | |||
GenBank mRNA | AF019085.1 | - | 4 | 526 | ||
AK302758.1 | - | 3 | 372 | |||
AK303008.1 | - | 4 | 516 | |||
AK316442.1 | - | 3 | 435 | |||
AY338951.1 | - | 5 | 837 | |||
BC005281.1 | - | 4 | 494 | |||
BC017582.1 | - | 4 | 569 | |||
BC047900.1 | - | 4 | 533 | |||
BC062700.1 | - | 4 | 528 | |||
BX648135.1 | - | 4 | 859 | |||
DB072781.1 | - | 4 | 519 |