Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322243.1 | NP_001309172.1 | 8 | 1377 | ||
NM_001728.3 | NP_001719.2 | 9 | 1729 | |||
NM_198589.2 | NP_940991.1 | 8 | 1381 | |||
NM_198590.2 | NP_940992.1 | 8 | 1436 | |||
NM_198591.2 | NP_940993.1 | 7 | 1279 | |||
XM_017027173.1 | XP_016882662.1 | 9 | 1614 | |||
GenBank mRNA | AF548371.1 | - | 9 | 1608 | ||
AK095912.1 | - | 7 | 1797 | |||
AK296858.1 | - | 6 | 894 | |||
AK302231.1 | - | 8 | 1249 | |||
AY358113.1 | - | 9 | 1600 | |||
BC009040.2 | - | 8 | 1244 | 3867352 | ||
D45131.1 | - | 8 | 1228 | |||
GU557064.1 | - | 8 | 1403 | |||
GU557065.1 | - | 7 | 1246 | |||
L10240.1 | - | 8 | 1214 | |||
L20471.1 | - | 8 | 1245 | |||
M87879.1 | - | 8 | 1222 | |||
X64364.1 | - | 8 | 1257 |