Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316963.1 | NP_001303892.1 | 3 | 582 | ||
NM_001729.3 | NP_001720.1 | 3 | 582 | |||
XM_011532211.1 | XP_011530513.1 | 3 | 563 | |||
GenBank mRNA | BC011618.2 | - | 3 | 373 | 3952444 | |
BI552049.1 | - | 3 | 399 | |||
KU177946.1 | - | 3 | 221 | |||
KU177947.1 | - | 3 | 221 | |||
S55606.1 | - | 3 | 515 |