Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5AR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001736.3 | NP_001727.1 | 2 | 1571 | ||
XM_005259190.4 | XP_005259247.1 | 2 | 1544 | |||
GenBank mRNA | BC008982.1 | - | 2 | 1597 | 4177114 | |
M62505.1 | - | 2 | 1544 |
Set Membership: |
G-protein coupled receptor |