Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001156.4 | NP_001147.1 | 9 | 944 | ||
NM_001320879.1 | NP_001307808.1 | 8 | 889 | |||
NM_001320880.1 | NP_001307809.1 | 9 | 955 | |||
NM_004034.3 | NP_004025.1 | 10 | 1010 | |||
XM_017016162.1 | XP_016871651.1 | 10 | 1046 | |||
XM_017016163.1 | XP_016871652.1 | 9 | 980 | |||
GenBank mRNA | AB062429.1 | - | 9 | 902 | ||
AK222552.1 | - | 9 | 912 | |||
AK295319.1 | - | 9 | 821 | |||
AK300086.1 | - | 7 | 711 | |||
AK301681.1 | - | 9 | 698 | |||
AK307772.1 | - | 8 | 762 | |||
AK309141.1 | - | 9 | 706 | |||
AK312447.1 | - | 10 | 983 | |||
AK313024.1 | - | 9 | 887 | |||
BC001322.1 | - | 2 | 195 | |||
BC002632.2 | - | 9 | 904 | 3610541 | ||
BT007187.1 | - | 8 | 869 | |||
BX361177.2 | - | 8 | 855 | |||
CR407686.1 | - | 8 | 869 | |||
J04543.1 | - | 9 | 929 |