Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3AR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326475.1 | NP_001313404.1 | 2 | 1914 | ||
NM_001326477.1 | NP_001313406.1 | 2 | 1974 | |||
NM_004054.3 | NP_004045.1 | 2 | 1917 | |||
GenBank mRNA | BC020742.1 | - | 2 | 1805 | 4690283 | |
U28488.1 | - | 2 | 1794 |
Set Membership: |
G-protein coupled receptor |