Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003954.2 | NP_001003954.1 | 5, 6 | 573 | ||
NM_004306.3 | NP_004297.2 | 4, 5 | 450 | |||
GenBank mRNA | AJ306450.1 | - | 5, 6 | 549 | ||
AK223374.1 | - | 4, 5 | 449 | |||
AK290281.1 | - | 5, 6 | 559 | |||
BC125158.1 | - | 4, 5 | 420 | 40120955 | ||
CR541838.1 | - | 3, 4 | 358 | |||
Z11502.1 | - | 4, 5 | 437 |