Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004455.2 | NP_004446.2 | 1 | 1213 | ||
XM_005245779.3 | XP_005245836.1 | 1 | 1244 | |||
XM_017000650.1 | XP_016856139.1 | 1 | 482 | |||
GenBank mRNA | BC065528.1 | - | 1 | 1059 | 6150218 | |
U67191.1 | - | 1 | 1212 |