Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004326.3 | NP_004317.2 | 10 | 4919 | ||
XM_005272971.4 | XP_005273028.1 | 10 | 4924 | |||
XM_005277417.3 | XP_005277474.1 | 10 | 4912 | |||
XM_005277418.3 | XP_005277475.1 | 10 | 4804 | |||
XM_017002026.1 | XP_016857515.1 | 10 | 4702 | |||
GenBank mRNA | BC116451.1 | - | 7 | 4314 | 7961941 | |
Y13620.1 | - | 10 | 5006 |