Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004327.3 | NP_004318.3 | 12 | 3138 | ||
NM_021574.2 | NP_067585.2 | 12 | 3138 | |||
GenBank mRNA | AB209991.1 | - | 12 | 2887 | ||
AK128501.1 | - | 4 | 1195 | |||
AK303241.1 | - | 12 | 1572 | |||
BC066122.1 | - | 12 | 2969 | |||
BQ422888.1 | - | 3 | 264 | |||
X02596.1 | - | 12 | 3030 | |||
Y00661.1 | - | 12 | 3138 |