Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004331.2 | NP_004322.1 | 6 | 1092 | ||
XM_005273617.3 | XP_005273674.1 | 6 | 996 | |||
GenBank mRNA | AB004788.1 | - | 6 | 1013 | ||
AF060922.1 | - | 6 | 1047 | |||
AF079221.1 | - | 6 | 1049 | |||
AF452712.2 | - | 6 | 1065 | |||
AF536326.1 | - | 6 | 1024 | |||
AK300727.1 | - | 6 | 885 | |||
AK307195.1 | - | 4 | 788 | |||
AK316315.1 | - | 6 | 996 | |||
AL132665.1 | - | 6 | 1066 | |||
BC001559.1 | - | 6 | 992 | 3459327 | ||
BC009603.1 | - | 6 | 1060 | 4134765 |