Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278616.1 | NP_001265545.1 | 7 | 756 | ||
NM_001278617.1 | NP_001265546.1 | 8 | 816 | |||
NM_004336.4 | NP_004327.1 | 8 | 816 | |||
GenBank mRNA | AF043294.2 | - | 8 | 736 | ||
AF046078.1 | - | 8 | 753 | |||
AF047471.1 | - | 8 | 739 | |||
AF053305.1 | - | 8 | 767 | |||
AK023540.1 | - | 8 | 772 | |||
AK225317.1 | - | 8 | 772 | |||
AK291221.1 | - | 8 | 769 | |||
AK302418.1 | - | 7 | 712 | |||
BC017038.1 | - | 8 | 737 | |||
BC028201.1 | - | 8 | 737 | 5205385 |