Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278616.1 | NP_001265545.1 | 7 | 685 | ||
NM_001278617.1 | NP_001265546.1 | 8 | 745 | |||
NM_004336.4 | NP_004327.1 | 8 | 745 | |||
GenBank mRNA | AF043294.2 | - | 8 | 665 | ||
AF046078.1 | - | 8 | 682 | |||
AF047471.1 | - | 8 | 668 | |||
AF053305.1 | - | 8 | 696 | |||
AK023540.1 | - | 8 | 701 | |||
AK225317.1 | - | 8 | 701 | |||
AK291221.1 | - | 8 | 698 | |||
AK302418.1 | - | 7 | 641 | |||
BC017038.1 | - | 8 | 666 | |||
BC028201.1 | - | 8 | 666 | 5205385 |