Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286822.1 | NP_001273751.1 | 3 | 483 | ||
NM_004339.3 | NP_004330.1 | 6 | 811 | |||
NR_104597.1 | 5 | 702 | ||||
GenBank mRNA | AF149785.1 | - | 6 | 749 | ||
AK094882.1 | - | 7 | 516 | |||
AK095586.1 | - | 1 | 1111 | |||
AK290139.1 | - | 6 | 561 | |||
AK293318.1 | - | 3 | 212 | |||
AK298557.1 | - | 3 | 271 | |||
AK299204.1 | - | 4 | 370 | |||
AK315626.1 | - | 6 | 626 | |||
BC000415.2 | - | 6 | 568 | 2961420 | ||
BC012858.1 | - | 6 | 594 | 3857505 | ||
BC019295.1 | - | 6 | 558 | 2823814 | ||
BC020983.1 | - | 6 | 575 | 3851447 | ||
BC031097.1 | - | 6 | 589 | 5288388 | ||
BC034250.1 | - | 6 | 583 | 3862830 | ||
CN295711.1 | - | 5 | 493 | |||
L48984.1 | - | 3 | 209 | |||
Z50022.1 | - | 6 | 617 |