Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318144.1 | NP_001305073.1 | 5 | 1571 | ||
NM_004925.4 | NP_004916.1 | 6 | 1789 | |||
GenBank mRNA | AK095363.1 | - | 3 | 2831 | ||
BC012988.2 | - | 1 | 449 | |||
BC013566.1 | - | 6 | 1723 | 3877096 |