Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005370.4 | NP_005361.2 | 3 | 435 | ||
XM_011528020.2 | XP_011526322.1 | 3 | 223 | |||
GenBank mRNA | AB209767.1 | - | 3 | 200 | ||
AF498943.1 | - | 3 | 213 | |||
AK025165.1 | - | 3 | 458 | |||
AK293676.1 | - | 3 | 218 | |||
BC002977.1 | - | 3 | 289 | 3547214 | ||
BT007184.1 | - | 3 | 213 | |||
CR536583.1 | - | 3 | 213 | |||
CR542274.1 | - | 3 | 213 | |||
X56741.1 | - | 3 | 223 |