Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164189.1 | NP_001157661.1 | 8 | 1702 | ||
NM_001164194.1 | NP_001157666.1 | 8 | 1669 | |||
NM_005817.4 | NP_005808.3 | 8 | 1705 | |||
GenBank mRNA | AF051315.1 | - | 4 | 993 | ||
AF055574.1 | - | 8 | 1581 | |||
AF057140.1 | - | 8 | 1602 | |||
AK122671.1 | - | 8 | 1556 | |||
AK225045.1 | - | 8 | 1571 | |||
AK225400.1 | - | 8 | 1586 | |||
AK226088.1 | - | 8 | 1583 | |||
AK291104.1 | - | 8 | 1662 | |||
BC001590.2 | - | 8 | 1578 | 2987965 | ||
BC005818.2 | - | 8 | 1594 | 3833411 | ||
BC007566.1 | - | 8 | 1594 | 3028104 | ||
BC019278.1 | - | 8 | 1594 | 2905275 |