Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005892.3 | NP_005883.2 | 25 | 3364 | ||
XM_006722062.3 | XP_006722125.1 | 26 | 3579 | |||
XM_006722063.3 | XP_006722126.1 | 25 | 3559 | |||
XM_006722064.3 | XP_006722127.1 | 26 | 3549 | |||
XM_006722065.3 | XP_006722128.1 | 25 | 3529 | |||
XM_006722066.3 | XP_006722129.1 | 26 | 3575 | |||
XM_006722069.2 | XP_006722132.1 | 25 | 3476 | |||
XM_006722070.3 | XP_006722133.1 | 25 | 3555 | |||
XM_011525179.2 | XP_011523481.1 | 26 | 3579 | |||
XM_011525180.2 | XP_011523482.1 | 26 | 3271 | |||
XM_011525182.2 | XP_011523484.1 | 26 | 3576 | |||
GenBank mRNA | AF432213.1 | - | 22 | 2838 | ||
AJ008112.1 | - | 10 | 1280 | |||
AK131091.1 | - | 16 | 4051 | |||
AY278319.1 | - | 25 | 3364 | |||
BC001710.1 | - | 10 | 1268 | |||
BC009000.2 | - | 5 | 545 | |||
BC021906.1 | - | 13 | 1872 | |||
BC073988.1 | - | 13 | 1912 | |||
BC111564.1 | - | 18 | 2190 | 40008718 | ||
CR456759.1 | - | 10 | 1241 | |||
FJ534522.1 | - | 9 | 1163 |