Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006015.4 | NP_006006.3 | 1 | 1011 | ||
NM_139135.2 | NP_624361.1 | 1 | 1011 | |||
GenBank mRNA | AF231056.1 | - | 1 | 1016 |