Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAGLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006133.2 | NP_006124.1 | 17 | 1884 | ||
XM_017018238.1 | XP_016873727.1 | 18 | 2211 | |||
XM_017018239.1 | XP_016873728.1 | 18 | 2116 | |||
XM_017018240.1 | XP_016873729.1 | 19 | 2333 | |||
GenBank mRNA | AB014559.2 | - | 17 | 1879 | ||
AY275377.1 | - | 16 | 1768 | |||
BC150176.1 | - | 17 | 1884 | 40148003 | ||
BC150195.1 | - | 17 | 1884 | 40148734 | ||
BC152453.1 | - | 17 | 1879 | 8862586 |