Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006134.6 | NP_006125.2 | 4 | 481 | ||
NR_040016.1 | 4 | 481 | ||||
XM_011529746.1 | XP_011528048.1 | 4 | 479 | |||
GenBank mRNA | AF045606.2 | - | 4 | 426 | ||
AF086280.1 | - | 3 | 315 | |||
AK309977.1 | - | 4 | 408 | |||
AK310701.1 | - | 4 | 337 | |||
AK311870.1 | - | 3 | 301 | |||
AY358634.1 | - | 4 | 405 | |||
AY820138.1 | - | 3 | 268 | |||
BC000569.2 | - | 4 | 397 | 3342648 | ||
DA286739.1 | - | 4 | 482 | |||
DQ286229.1 | - | 3 | 268 |