Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181575.4 | NP_853553.1 | 7 | 982 | ||
NR_126510.1 | 7 | 982 | ||||
NR_126511.1 | 6 | 1178 | ||||
XM_005264392.3 | XP_005264449.1 | 6 | 927 | |||
GenBank mRNA | AF100746.1 | - | 7 | 755 | ||
AF100753.1 | - | 7 | 782 | |||
AF100754.1 | - | 6 | 980 | |||
AF165515.1 | - | 7 | 762 | |||
AK023983.1 | - | 7 | 760 | |||
AK055566.1 | - | 4 | 1513 | |||
AK293121.1 | - | 7 | 801 | |||
AK304886.1 | - | 6 | 924 | |||
AK308231.1 | - | 6 | 981 | |||
AK310159.1 | - | 6 | 980 | |||
BC001658.1 | - | 7 | 845 | 3050204 | ||
BC033646.2 | - | 7 | 797 | |||
CR749661.1 | - | 6 | 853 |