Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199397.1 | NP_001186326.1 | 26 | 2797 | ||
NM_001199398.1 | NP_001186327.1 | 25 | 2665 | |||
NM_001199399.1 | NP_001186328.1 | 23 | 2506 | |||
NM_001199400.1 | NP_001186329.1 | 23 | 2659 | |||
NM_012224.2 | NP_036356.1 | 24 | 2791 | |||
XM_006714228.1 | XP_006714291.1 | 26 | 2799 | |||
XM_011532003.1 | XP_011530305.1 | 25 | 2715 | |||
XM_011532004.1 | XP_011530306.1 | 24 | 2583 | |||
XM_017008249.1 | XP_016863738.1 | 25 | 2701 | |||
XM_017008250.1 | XP_016863739.1 | 24 | 2819 | |||
XM_017008251.1 | XP_016863740.1 | 24 | 2617 | |||
XM_017008252.1 | XP_016863741.1 | 22 | 2637 | |||
XM_017008253.1 | XP_016863742.1 | 25 | 2647 | |||
XM_017008254.1 | XP_016863743.1 | 18 | 1757 | |||
GenBank mRNA | AB067488.1 | - | 24 | 2627 | ||
AF155113.1 | - | 8 | 675 | |||
BC114491.1 | - | 23 | 2266 | 40082305 | ||
CR933642.1 | - | 26 | 2755 | |||
CR936672.1 | - | 23 | 2617 |