Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258328.2 | NP_001245257.1 | 13 | 1676 | ||
NM_001282334.1 | NP_001269263.1 | 3 | 1563 | |||
NM_001282335.1 | NP_001269264.1 | 12 | 1242 | |||
NM_001321232.1 | NP_001308161.1 | 13 | 1612 | |||
NM_012292.4 | NP_036424.2 | 13 | 1765 | |||
XM_006722713.1 | XP_006722776.1 | 13 | 1765 | |||
XM_011527858.1 | XP_011526160.1 | 13 | 1612 | |||
XM_011527860.2 | XP_011526162.1 | 4 | 585 | |||
GenBank mRNA | AK122776.1 | - | 3 | 754 | ||
AK128097.1 | - | 12 | 1239 | |||
AK300341.1 | - | 13 | 1668 | |||
AK304271.1 | - | 13 | 1636 | |||
AK304765.1 | - | 12 | 1283 | |||
BC035564.1 | - | 13 | 1510 | |||
BC065223.1 | - | 13 | 1757 | 5751491 | ||
BU838153.1 | - | 3 | 225 | |||
D86976.1 | - | 13 | 1613 |