Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258328.2 | NP_001245257.1 | 23 | 4153 | ||
NM_001282334.1 | NP_001269263.1 | 13 | 4040 | |||
NM_001282335.1 | NP_001269264.1 | 22 | 3719 | |||
NM_001321232.1 | NP_001308161.1 | 23 | 4089 | |||
NM_012292.4 | NP_036424.2 | 23 | 4242 | |||
XM_006722713.1 | XP_006722776.1 | 22 | 4326 | |||
XM_011527858.1 | XP_011526160.1 | 22 | 4173 | |||
XM_011527860.2 | XP_011526162.1 | 14 | 3062 | |||
GenBank mRNA | AK122776.1 | - | 13 | 3231 | ||
AK128097.1 | - | 22 | 3716 | |||
BC035564.1 | - | 23 | 3987 | |||
BC065223.1 | - | 23 | 4234 | 5751491 | ||
D86976.1 | - | 23 | 4090 |