Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242837.1 | NP_001229766.1 | 3 | 464 | ||
NM_012305.3 | NP_036437.1 | 3 | 464 | |||
XM_011519929.1 | XP_011518231.1 | 3 | 221 | |||
XM_011519930.2 | XP_011518232.1 | 3 | 425 | |||
GenBank mRNA | AB020706.2 | - | 3 | 251 | ||
AK001986.1 | - | 3 | 393 | |||
AK027891.1 | - | 3 | 373 | |||
AK223558.1 | - | 3 | 392 | |||
AK293763.1 | - | 3 | 323 | |||
AK299364.1 | - | 3 | 242 | |||
BC006155.1 | - | 3 | 392 | 3627239 | ||
BG705558.1 | - | 3 | 432 | |||
BI462785.1 | - | 3 | 442 |