Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127392.2 | NP_001120864.1 | 14 | 2042 | ||
NM_013279.3 | NP_037411.1 | 14 | 1942 | |||
XM_005274222.1 | XP_005274279.1 | 14 | 2030 | |||
XM_005274223.1 | XP_005274280.1 | 14 | 2030 | |||
XM_005274224.1 | XP_005274281.1 | 14 | 2030 | |||
XM_005274225.1 | XP_005274282.1 | 14 | 2030 | |||
XM_005274226.1 | XP_005274283.1 | 14 | 2030 | |||
XM_005274227.1 | XP_005274284.1 | 14 | 2030 | |||
XM_005274228.1 | XP_005274285.1 | 14 | 1877 | |||
XM_011545234.2 | XP_011543536.1 | 14 | 1916 | |||
GenBank mRNA | AB023171.2 | - | 14 | 2030 | ||
AF038536.1 | - | 5 | 510 | |||
AF086762.1 | - | 14 | 1942 | |||
AK295013.1 | - | 2 | 271 |