Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014470.3 | NP_055285.1 | 2 | 297 | ||
GenBank mRNA | AB040147.1 | - | 2 | 275 | ||
AF498967.1 | - | 2 | 194 | |||
AK292762.1 | - | 2 | 297 | |||
BC026356.1 | - | 2 | 297 | 4793442 | ||
Y07923.1 | - | 2 | 194 |