Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257962.1 | NP_001244891.1 | 3, 4 | 457 | ||
NM_014476.5 | NP_055291.2 | 3, 4 | 457 | |||
GenBank mRNA | AF002280.1 | - | 3, 4 | 379 | ||
AF039018.1 | - | 3, 4 | 403 | |||
AK222966.1 | - | 3, 4 | 361 | |||
AK313253.1 | - | 3, 4 | 406 | |||
C03422.1 | - | 3, 4 | 283 |