Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301132.1 | NP_001288061.1 | 7 | 2675 | ||
NM_014952.4 | NP_055767.3 | 7 | 2551 | |||
NR_125372.1 | 5 | 713 | ||||
XM_011521366.2 | XP_011519668.2 | 7 | 2675 | |||
XM_011521367.2 | XP_011519669.2 | 7 | 2677 | |||
XM_011521368.2 | XP_011519670.2 | 7 | 2665 | |||
XM_011521369.2 | XP_011519671.1 | 7 | 2529 | |||
GenBank mRNA | AB023162.1 | - | 7 | 2675 | ||
AL833923.1 | - | 6 | 1634 | |||
BC022782.2 | - | 7 | 2490 | 3903581 |