Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319245.1 | NP_001306174.1 | 4, 5 | 788 | ||
NM_001324320.1 | NP_001311249.1 | 4, 5 | 794 | |||
NM_015112.2 | NP_055927.2 | 4, 5 | 788 | |||
XM_017000755.1 | XP_016856244.1 | 2, 3 | 799 | |||
GenBank mRNA | AB047005.1 | - | 4, 5 | 788 | ||
AK055291.1 | - | 2, 3 | 799 | |||
BC065499.1 | - | 4, 5 | 762 | 5499830 |