Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083625.2 | NP_001077094.1 | 3 | 405 | ||
NM_001204056.1 | NP_001190985.1 | 3 | 503 | |||
NM_015208.4 | NP_056023.3 | 3 | 405 | |||
XM_005258092.3 | XP_005258149.1 | 3 | 538 | |||
XM_005258093.3 | XP_005258150.1 | 3 | 522 | |||
XM_005258095.2 | XP_005258152.1 | 3 | 377 | |||
XM_005258096.2 | XP_005258153.1 | 3 | 381 | |||
XM_011525638.2 | XP_011523940.1 | 3 | 377 | |||
XM_011525639.2 | XP_011523941.1 | 3 | 494 | |||
XM_017025661.1 | XP_016881150.1 | 3 | 506 | |||
XM_017025662.1 | XP_016881151.1 | 3 | 551 | |||
XM_017025663.1 | XP_016881152.1 | 3 | 490 | |||
XM_017025664.1 | XP_016881153.1 | 3 | 490 | |||
XM_017025665.1 | XP_016881154.1 | 3 | 524 | |||
XM_017025666.1 | XP_016881155.1 | 3 | 463 | |||
XM_017025667.1 | XP_016881156.1 | 3 | 319 | |||
XM_017025668.1 | XP_016881157.1 | 3 | 319 | |||
GenBank mRNA | AF317425.1 | - | 2 | 141 | ||
AK024808.1 | - | 3 | 355 | |||
AL834204.1 | - | 3 | 333 | |||
AY373757.1 | - | 3 | 355 | |||
AY533564.2 | - | 3 | 503 | |||
BC057225.1 | - | 3 | 354 | |||
BC070234.1 | - | 3 | 375 | |||
BP230422.1 | - | 3 | 405 | |||
BP255733.1 | - | 3 | 398 |